Learn More
Eurofins Discoverx PathHunter eXpress IL10RA/IL10RB Dimerization Assay, 2pl Kit. (93-0985E3CP0M)
Catalog No. 502001749
Encompass
The PathHunter IL10RA/IL10RB Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-10) induced dimerization of IL10RA/IL10RB receptors. This product uses EFC technology. The cell line included overexpresses IL10RA target protein fused to PK alongside IL10RB target protein fused to EA. Upon ligand-induced dimerization of IL10RA/IL10RB, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1749 Supplier Eurofins Discoverx Supplier No. 930985E3CP0M
The PathHunter IL10RA/IL10RB Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-10) induced dimerization of IL10RA/IL10RB receptors. This product uses EFC technology. The cell line included overexpresses IL10RA target protein fused to PK alongside IL10RB target protein fused to EA. Upon ligand-induced dimerization of IL10RA/IL10RB, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.