Learn More
Eurofins Discoverx PathHunter eXpress IL21R/IL2RG Dimerization Assay, 2pl Kit. (93-1035E3CP0M)
Catalog No. 502001898
Encompass
The PathHunter IL21R/IL2RG Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-21) induced dimerization of IL21R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL21RG target protein fused to PK alongside IL2R target protein fused to EA. Upon ligand-induced dimerization of IL21R/IL2RG, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1898 Supplier Eurofins Discoverx Supplier No. 931035E3CP0M
Add to Cart
Add to Cart
The PathHunter IL21R/IL2RG Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-21) induced dimerization of IL21R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL21RG target protein fused to PK alongside IL2R target protein fused to EA. Upon ligand-induced dimerization of IL21R/IL2RG, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.