Learn More
Eurofins Discoverx PathHunter eXpress IL2RB/IL2RG/IL2RA Dimerization Assay, 10pl Kit. (93-1003E3CP0L)
Catalog No. 502001812
Encompass
The PathHunter IL2RB/IL2RG/IL2RA Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-2) induced dimerization of IL2RB/IL2RG/IL2RA receptors. This product uses EFC technology. The cell line included overexpresses IL2RB target protein fused to PK alongside IL2RG target protein fused to EA and untagged ILR2A. Upon ligand-induced dimerization of IL2RB/IL2RG/IL2RA, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1812 Supplier Eurofins Discoverx Supplier No. 931003E3CP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL2RB/IL2RG/IL2RA Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-2) induced dimerization of IL2RB/IL2RG/IL2RA receptors. This product uses EFC technology. The cell line included overexpresses IL2RB target protein fused to PK alongside IL2RG target protein fused to EA and untagged ILR2A. Upon ligand-induced dimerization of IL2RB/IL2RG/IL2RA, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.