Learn More
Eurofins Discoverx PathHunter eXpress IL4R/IL13RA1 Dimerization Assay, HEK293,10pl Kit. (93-1001E1CP0L)
Catalog No. 502001800
Encompass
The PathHunter IL4R/IL13RA1 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-13) induced dimerization of IL4R/IL13RA1 receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL13RA1 target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL13RA1, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1800 Supplier Eurofins Discoverx Supplier No. 931001E1CP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL4R/IL13RA1 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-13) induced dimerization of IL4R/IL13RA1 receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL13RA1 target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL13RA1, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.