Learn More
Eurofins Discoverx PathHunter eXpress IL4R/IL2RG Dimerization Assay, 2pl Kit. (93-0988E3CP5M)
Supplier:  Eurofins Discoverx 930988E3CP5M
Encompass
The PathHunter IL4R/IL2RG Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-4) induced dimerization of IL4R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL2RG, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1757