Learn More
Eurofins Discoverx PathHunter eXpress IL5RA/CSF2RB Dimerization Assay, 10pl Kit. (93-0972E3CP0L)
Catalog No. 502001714
Encompass
The PathHunter IL5RA/CSF2RB Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-5) induced dimerization of IL5RA/CSF2RB receptors. This product uses EFC technology. The cell line included overexpresses CSF2RB target protein fused to PK alongside IL5RA target protein fused to EA. Upon ligand-induced dimerization of IL5RA/CSF2RB, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1714 Supplier Eurofins Discoverx Supplier No. 930972E3CP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL5RA/CSF2RB Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-5) induced dimerization of IL5RA/CSF2RB receptors. This product uses EFC technology. The cell line included overexpresses CSF2RB target protein fused to PK alongside IL5RA target protein fused to EA. Upon ligand-induced dimerization of IL5RA/CSF2RB, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.