Learn More
Eurofins Discoverx PathHunter eXpress IL7R/CRLF2 Dimerization Assay, 2pl Kit. (93-1019E13CP25M)
Catalog No. 502001863
Encompass
The PathHunter IL7R/CRLF2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. TSLP) induced dimerization of IL7R/CRLF2 receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside CRLF2 target protein fused to EA. Upon ligand-induced dimerization of IL7R/CRLF2, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1863 Supplier Eurofins Discoverx Supplier No. 931019E13CP25M
Add to Cart
Add to Cart
The PathHunter IL7R/CRLF2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. TSLP) induced dimerization of IL7R/CRLF2 receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside CRLF2 target protein fused to EA. Upon ligand-induced dimerization of IL7R/CRLF2, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.