Learn More
Eurofins Discoverx PathHunter eXpress IL7R/IL2RG Dimerization Assay, 10pl Kit. (93-0997E13CP0L)
Catalog No. 502001781
Encompass
The PathHunter IL7R/IL2RG Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-7) induced dimerization of IL7R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL7R/IL2RG, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1781 Supplier Eurofins Discoverx Supplier No. 930997E13CP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL7R/IL2RG Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-7) induced dimerization of IL7R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL7R/IL2RG, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.