Learn More
Eurofins Discoverx PathHunter eXpress Keap1-NRF2 Nuclear Translocation Assay, 2-plate Kit. (93-0821E3CP0M)
Catalog No. 502001357
Encompass
The PathHunter eXpress Keap1-NRF2 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the Keap1-NRF2 pathway. This kit assesses ligand (e.g. methyl CDDO) based pathway activation via detection of resulting NRF2 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged NRF2 and nuclear localized EA alongside untagged Keap1. Upon pathway activation, induced nuclear translocation of NRF2 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1357 Supplier Eurofins Discoverx Supplier No. 930821E3CP0M
The PathHunter eXpress Keap1-NRF2 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the Keap1-NRF2 pathway. This kit assesses ligand (e.g. methyl CDDO) based pathway activation via detection of resulting NRF2 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged NRF2 and nuclear localized EA alongside untagged Keap1. Upon pathway activation, induced nuclear translocation of NRF2 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.