Learn More
Eurofins Discoverx PathHunter eXpress mADCYAP1R1 (PAC1) CHO-K1 β-Arrestin GPCR Assay, 10pl Kit. (93-0594E2MCP0L)
Catalog No. 502000914
Encompass
The PathHunter mADCYAP1R1 (PAC1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. PACAP) based activation of mADCYAP1R1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mADCYAP1R1 and EA-tagged β-Arrestin-2. Activation of mADCYAP1R1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0914 Supplier Eurofins Discoverx Supplier No. 930594E2MCP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter mADCYAP1R1 (PAC1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. PACAP) based activation of mADCYAP1R1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mADCYAP1R1 and EA-tagged β-Arrestin-2. Activation of mADCYAP1R1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.