Learn More
Eurofins Discoverx PathHunter eXpress mCCR7 CHO-K1 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0528PE2MCP0M)
Catalog No. 502000726
Encompass
The PathHunter mCCR7 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CCL19/MIP-3β) based activation of mCCR7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCCR7 and EA-tagged β-Arrestin-2. Activation of mCCR7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0726 Supplier Eurofins Discoverx Supplier No. 930528PE2MCP0M
The PathHunter mCCR7 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CCL19/MIP-3β) based activation of mCCR7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCCR7 and EA-tagged β-Arrestin-2. Activation of mCCR7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.