Learn More
Eurofins Discoverx PathHunter eXpress mCCR9 CHO-K1 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0734E2MCP2M)
Catalog No. 502001209
Encompass
The PathHunter mCCR9 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CCL25) based activation of mCCR9 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCCR9 and EA-tagged β-Arrestin-2. Activation of mCCR9 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1209 Supplier Eurofins Discoverx Supplier No. 930734E2MCP2M
The PathHunter mCCR9 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CCL25) based activation of mCCR9 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCCR9 and EA-tagged β-Arrestin-2. Activation of mCCR9 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.