Learn More
Eurofins Discoverx PathHunter eXpress mCXCR4 U2OS β-Arrestin GPCR Assay, 10-Plate Kit. (93-0481E3MCP5L)
Catalog No. 502000628
Encompass
The PathHunter mCXCR4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CXCL12/SDF-1α) based activation of mCXCR4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCXCR4 and EA-tagged β-Arrestin-2. Activation of mCXCR4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0628 Supplier Eurofins Discoverx Supplier No. 930481E3MCP5L
Add to Cart
Add to Cart
Request a Quote
The PathHunter mCXCR4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CXCL12/SDF-1α) based activation of mCXCR4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCXCR4 and EA-tagged β-Arrestin-2. Activation of mCXCR4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.