Learn More
Eurofins Discoverx PathHunter eXpress mGCGR CHO-K1 β-Arrestin-1 GPCR Assay, 10pl Kit. (93-0973E2MCP2L)
Catalog No. 502001718
Encompass
The PathHunter mGCGR (ARR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. glucagon) based activation of mGCGR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGCGR and EA-tagged β-Arrestin-1. Activation of mGCGR stimulates the recruitment of β-Arrestin-1 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1718 Supplier Eurofins Discoverx Supplier No. 930973E2MCP2L
Add to Cart
Add to Cart
Request a Quote
The PathHunter mGCGR (ARR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. glucagon) based activation of mGCGR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGCGR and EA-tagged β-Arrestin-1. Activation of mGCGR stimulates the recruitment of β-Arrestin-1 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.