Learn More
Eurofins Discoverx PathHunter eXpress mGLP1R CHO-K1 β-Arrestin-1 GPCR Assay, 10pl Kit. (93-0974E2MCP2L)
Catalog No. 502001722
Encompass
The PathHunter mGLP1R (ARR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. exendin-4) based activation of mGLP1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGLP1R and EA-tagged β-Arrestin-1. Activation of mGLP1R stimulates the recruitment of β-Arrestin-1 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1722 Supplier Eurofins Discoverx Supplier No. 930974E2MCP2L
Add to Cart
Add to Cart
Request a Quote
The PathHunter mGLP1R (ARR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. exendin-4) based activation of mGLP1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGLP1R and EA-tagged β-Arrestin-1. Activation of mGLP1R stimulates the recruitment of β-Arrestin-1 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.