Learn More
Eurofins Discoverx PathHunter eXpress mPTGER4 DLD1 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0982E13MCP25L)
Catalog No. 502001742
Encompass
The PathHunter mPTGER4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin E2) based activation of mPTGER4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mPTGER4 and EA-tagged β-Arrestin-2. Activation of mPTGER4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1742 Supplier Eurofins Discoverx Supplier No. 930982E13MCP25L
Add to Cart
Add to Cart
Request a Quote
The PathHunter mPTGER4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin E2) based activation of mPTGER4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mPTGER4 and EA-tagged β-Arrestin-2. Activation of mPTGER4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.