Learn More
Eurofins Discoverx PathHunter eXpress mRXFP4 CHO-K1 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0929E2MCP2M)
Catalog No. 502001590
Encompass
The PathHunter mRXFP4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. insulin-like 5, relaxin-3) based activation of mRXFP4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mRXFP4 and EA-tagged β-Arrestin-2. Activation of mRXFP4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1590 Supplier Eurofins Discoverx Supplier No. 930929E2MCP2M
The PathHunter mRXFP4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. insulin-like 5, relaxin-3) based activation of mRXFP4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mRXFP4 and EA-tagged β-Arrestin-2. Activation of mRXFP4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.