Learn More
Eurofins Discoverx PathHunter eXpress mUTR2 CHO-K1 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0525E2MCP0M)
Catalog No. 502000714
Encompass
The PathHunter mUTR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. urotensin II) based activation of mUTR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mUTR2 and EA-tagged β-Arrestin-2. Activation of mUTR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0714 Supplier Eurofins Discoverx Supplier No. 930525E2MCP0M
The PathHunter mUTR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. urotensin II) based activation of mUTR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mUTR2 and EA-tagged β-Arrestin-2. Activation of mUTR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.