Learn More
Eurofins Discoverx cAMP Hunter eXpress OPRD1 CHO-K1 GPCR Assay, 10-Plate Kit. (95-0108E2CP2L)
Catalog No. 502003171
Encompass
The cAMP Hunter OPRD1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. DADLE, DPDPE) based activation of OPRD1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type OPRD1, a naturally Gi/Go coupled GPCR. Upon activation of OPRD1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3171 Supplier Eurofins Discoverx Supplier No. 950108E2CP2L
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter OPRD1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. DADLE, DPDPE) based activation of OPRD1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type OPRD1, a naturally Gi/Go coupled GPCR. Upon activation of OPRD1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.