Learn More
Eurofins Discoverx cAMP Hunter eXpress OXER1 CHO-K1 GPCR Assay, 2-Plate Kit. (95-0089E2CP2M)
Catalog No. 502002910
Encompass
The cAMP Hunter OXER1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. 5-oxo-ETE) based activation of OXER1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type OXER1, a naturally Gi/Go coupled GPCR. Upon activation of OXER1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2910 Supplier Eurofins Discoverx Supplier No. 950089E2CP2M
The cAMP Hunter OXER1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. 5-oxo-ETE) based activation of OXER1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type OXER1, a naturally Gi/Go coupled GPCR. Upon activation of OXER1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.