Learn More
Charter Medical 190 mL Cell Expansion Bio-Containers, 12/pk
Catalog No. NC1783640
Encompass
190 mL fill volume at 1 cm thick cell expansion bag with two leads and female luers, and dockable fill line with sampling valve, 12/pk.
This item is not returnable. View return policy
Catalog No. NC1783640 Supplier Charter Medical Supplier No. EXP500
190 mL fill volume at 1 cm thick cell expansion bag with two leads and female luers, and dockable fill line with sampling valve, 12/pk.