Learn More
Charter Medical Exp-Pak, 30mL fill volume, 24/Case
Catalog No. NC1783629
Encompass
30mL fill volume at 1cm thick cell expansion bag with two leads and female luers, and dockable fill line with sampling valve
This item is not returnable. View return policy
Catalog No. NC1783629 Supplier Charter Medical Supplier No. EXP50
30mL fill volume at 1cm thick cell expansion bag with two leads and female luers, and dockable fill line with sampling valve