Learn More
MSC Karcher Suction Hose, PU Plus, 5 m, 50-40 mm
Catalog No. 502837591
Encompass
This PU Plus suction hose is designed for industrial vacuum applications. It is 5 meters long, features a durable PU Plus material construction, and transitions in diameter from 50 mm to 40 mm. It is suitable for use in environments with temperatures up to 140°F.
  • Suction hose, PU Plus
  • Constructed from durable PU Plus material
  • 5 m length
  • 50 mm to 40 mm diameter
  • Withstands temperatures up to 140°F
  • Electrically conductive
Catalog No. 50-283-7591 Supplier MSC Supplier No. 48041867
This PU Plus suction hose is designed for industrial vacuum applications. It is 5 meters long, features a durable PU Plus material construction, and transitions in diameter from 50 mm to 40 mm. It is suitable for use in environments with temperatures up to 140°F.
  • Suction hose, PU Plus
  • Constructed from durable PU Plus material
  • 5 m length
  • 50 mm to 40 mm diameter
  • Withstands temperatures up to 140°F
  • Electrically conductive