Learn More
Eurofins Discoverx cAMP Hunter eXpress PTGDR CHO-K1 GPCR Assay, 10-Plate Kit. (95-0110E2CP2L)
Catalog No. 502003196
Encompass
The cAMP Hunter PTGDR eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin D2) based activation of PTGDR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTGDR, a naturally Gs coupled GPCR. Upon activation of PTGDR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3196 Supplier Eurofins Discoverx Supplier No. 950110E2CP2L
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter PTGDR eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin D2) based activation of PTGDR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTGDR, a naturally Gs coupled GPCR. Upon activation of PTGDR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.