Learn More
Eurofins Discoverx PathHunter eXpress PTGER4 HEK 293 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0304E1CP0M)
Catalog No. 502000217
Encompass
The PathHunter PTGER4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin E2) based activation of PTGER4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGER4 and EA-tagged β-Arrestin-2. Activation of PTGER4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0217 Supplier Eurofins Discoverx Supplier No. 930304E1CP0M
Add to Cart
Add to Cart
The PathHunter PTGER4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin E2) based activation of PTGER4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGER4 and EA-tagged β-Arrestin-2. Activation of PTGER4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.