Learn More
Eurofins Discoverx cAMP Hunter eXpress PTHR1 CHO-K1 GPCR Assay, 10-Plate Kit. (95-0118E2CP2L)
Catalog No. 502003264
Encompass
The cAMP Hunter PTHR1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. parathyroid hormone) based activation of PTHR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTHR1, a naturally Gs coupled GPCR. Upon activation of PTHR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3264 Supplier Eurofins Discoverx Supplier No. 950118E2CP2L
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter PTHR1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. parathyroid hormone) based activation of PTHR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTHR1, a naturally Gs coupled GPCR. Upon activation of PTHR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.