Learn More
Eurofins Discoverx cAMP Hunter eXpress PTHR2 CHO-K1 GPCR Assay, 2-Plate Kit. (95-0057E2CP2M)
Catalog No. 502002568
Encompass
The cAMP Hunter PTHR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. parathyroid hormone, TIP 39) based activation of PTHR2 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTHR2, a naturally Gs coupled GPCR. Upon activation of PTHR2, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2568 Supplier Eurofins Discoverx Supplier No. 950057E2CP2M
The cAMP Hunter PTHR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. parathyroid hormone, TIP 39) based activation of PTHR2 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTHR2, a naturally Gs coupled GPCR. Upon activation of PTHR2, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.