Learn More
Eurofins Discoverx PathHunter eXpress rCHRM4 CHO-K1 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0870E2RCP2L)
Catalog No. 502001453
Encompass
The PathHunter rCHRM4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. acetylcholine) based activation of rCHRM4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rCHRM4 and EA-tagged β-Arrestin-2. Activation of rCHRM4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1453 Supplier Eurofins Discoverx Supplier No. 930870E2RCP2L
Add to Cart
Add to Cart
Request a Quote
The PathHunter rCHRM4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. acetylcholine) based activation of rCHRM4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rCHRM4 and EA-tagged β-Arrestin-2. Activation of rCHRM4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.