Learn More
Eurofins Discoverx PathHunter eXpress rCRTH2 CHO-K1 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0580E2RCP2L)
Catalog No. 502000881
Encompass
The PathHunter rCRTH2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin D2) based activation of rCRTH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rCRTH2 and EA-tagged β-Arrestin-2. Activation of rCRTH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0881 Supplier Eurofins Discoverx Supplier No. 930580E2RCP2L
Add to Cart
Add to Cart
Request a Quote
The PathHunter rCRTH2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prostaglandin D2) based activation of rCRTH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rCRTH2 and EA-tagged β-Arrestin-2. Activation of rCRTH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.