Learn More
Eurofins Discoverx cAMP Hunter eXpress rDRD2L (Long Isoform) CHO-K1 GPCR Assay, 2pl Kit. (95-0130E2CP2M)
Catalog No. 502003591
Encompass
The cAMP Hunter rDRD2L eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. dopamine, pergolide) based activation of rDRD2L GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type rDRD2L, a naturally Gi/Go coupled GPCR. Upon activation of rDRD2L, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3591 Supplier Eurofins Discoverx Supplier No. 950130E2CP2M
The cAMP Hunter rDRD2L eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. dopamine, pergolide) based activation of rDRD2L GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type rDRD2L, a naturally Gi/Go coupled GPCR. Upon activation of rDRD2L, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.