Learn More
Eurofins Discoverx PathHunter eXpress rGPR120 CHO-K1 β-Arrestin GPCR Assay, 10pl Kit. (93-1031E2RCP0L)
Catalog No. 502001886
Encompass
The PathHunter rGPR120 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. free fatty acids) based activation of rGPR120 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rGPR120 and EA-tagged β-Arrestin-2. Activation of rGPR120 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1886 Supplier Eurofins Discoverx Supplier No. 931031E2RCP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter rGPR120 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. free fatty acids) based activation of rGPR120 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rGPR120 and EA-tagged β-Arrestin-2. Activation of rGPR120 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.