Learn More
Eurofins Discoverx PathHunter eXpress rGPR35 CHO-K1 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0887E2RCP2L)
Catalog No. 502001503
Encompass
The PathHunter rGPR35 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. zaprinast) based activation of rGPR35 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rGPR35 and EA-tagged β-Arrestin-2. Activation of rGPR35 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1503 Supplier Eurofins Discoverx Supplier No. 930887E2RCP2L
Add to Cart
Add to Cart
Request a Quote
The PathHunter rGPR35 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. zaprinast) based activation of rGPR35 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rGPR35 and EA-tagged β-Arrestin-2. Activation of rGPR35 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.