Learn More
Eurofins Discoverx PathHunter eXpress rHTR2C U2OS β-Arrestin GPCR Assay, 10-Plate Kit. (93-0915E3RCP0L)
Catalog No. 502001557
Encompass
The PathHunter rHTR2C eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. serotonin) based activation of rHTR2C GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rHTR2C and EA-tagged β-Arrestin-2. Activation of rHTR2C stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1557 Supplier Eurofins Discoverx Supplier No. 930915E3RCP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter rHTR2C eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. serotonin) based activation of rHTR2C GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rHTR2C and EA-tagged β-Arrestin-2. Activation of rHTR2C stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.