Learn More
Eurofins Discoverx PathHunter eXpress rhTrkA-rhP75 Functional Assay, 2pl Kit. (93-1009E3CP16M)
Catalog No. 502001830
Encompass
The PathHunter eXpress rhTrkA-rhP75 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated rhTrkA and rhP75 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged rhTrkA and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, rhP75 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the rhesus version of the protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1830 Supplier Eurofins Discoverx Supplier No. 931009E3CP16M
The PathHunter eXpress rhTrkA-rhP75 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated rhTrkA and rhP75 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged rhTrkA and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, rhP75 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the rhesus version of the protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.