Learn More
Eurofins Discoverx PathHunter eXpress rOPRM1 CHO-K1 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0494E2RCP2L)
Catalog No. 502000655
Encompass
The PathHunter rOPRM1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. DADLE) based activation of rOPRM1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rOPRM1 and EA-tagged β-Arrestin-2. Activation of rOPRM1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0655 Supplier Eurofins Discoverx Supplier No. 930494E2RCP2L
Add to Cart
Add to Cart
Request a Quote
The PathHunter rOPRM1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. DADLE) based activation of rOPRM1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rOPRM1 and EA-tagged β-Arrestin-2. Activation of rOPRM1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.