Learn More
Eurofins Discoverx PathHunter eXpress rPROKR2 CHO-K1 β-Arrestin GPCR Assay, 10pl Kit. (93-0482E2RCP0L)
Catalog No. 502000630
Encompass
The PathHunter rPROKR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prokineticin) based activation of rPROKR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rPROKR2 and EA-tagged β-Arrestin-2. Activation of rPROKR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0630 Supplier Eurofins Discoverx Supplier No. 930482E2RCP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter rPROKR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. prokineticin) based activation of rPROKR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rPROKR2 and EA-tagged β-Arrestin-2. Activation of rPROKR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.