Learn More
Eurofins Discoverx PathHunter eXpress TACR3 CHO-K1 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0501E2CP2M)
Catalog No. 502000664
Encompass
The PathHunter TACR3 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. substance P, senkitide) based activation of TACR3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TACR3 and EA-tagged β-Arrestin-2. Activation of TACR3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0664 Supplier Eurofins Discoverx Supplier No. 930501E2CP2M
The PathHunter TACR3 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. substance P, senkitide) based activation of TACR3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TACR3 and EA-tagged β-Arrestin-2. Activation of TACR3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.