Learn More
Eurofins Discoverx PathHunter eXpress TBXA2R U2OS β-Arrestin GPCR Assay, 10-Plate Kit. (93-0322E3CP5L)
Catalog No. 502000278
Encompass
The PathHunter TBXA2R eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. thromboxane A2, prostaglandins) based activation of TBXA2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TBXA2R and EA-tagged β-Arrestin-2. Activation of TBXA2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0278 Supplier Eurofins Discoverx Supplier No. 930322E3CP5L
Add to Cart
Add to Cart
Request a Quote
The PathHunter TBXA2R eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. thromboxane A2, prostaglandins) based activation of TBXA2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TBXA2R and EA-tagged β-Arrestin-2. Activation of TBXA2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.