Learn More
Eurofins Discoverx PathHunter eXpress TGFBR1/TGFBR2 Dimerization Assay, 2pl Kit. (93-0889E3CP5M)
Catalog No. 502001512
Encompass
The PathHunter TGFBR1/TGFBR2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. TGF-β1) induced dimerization of TGFBR1/TGFBR2 receptors. This product uses EFC technology. The cell line included overexpresses TGFBR1 target protein fused to PK alongside TGFBR2 target protein fused to EA. Upon ligand-induced dimerization of TGFBR1/TGFBR2, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1512 Supplier Eurofins Discoverx Supplier No. 930889E3CP5M
The PathHunter TGFBR1/TGFBR2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. TGF-β1) induced dimerization of TGFBR1/TGFBR2 receptors. This product uses EFC technology. The cell line included overexpresses TGFBR1 target protein fused to PK alongside TGFBR2 target protein fused to EA. Upon ligand-induced dimerization of TGFBR1/TGFBR2, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.