Learn More
Eurofins Discoverx PathHunter eXpress TORC2 Nuclear Translocation Assay, 10pl Kit. (93-0813E3CP19L)
Catalog No. 502001336
Encompass
The PathHunter eXpress TORC2 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the CRTC/CREB pathway. This kit assesses ligand (e.g. forskolin) based pathway activation via detection of resulting TORC2 (CRTC2) nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged TORC2 (CRTC2) and nuclear localized EA. Upon pathway activation, induced nuclear translocation of TORC2 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1336 Supplier Eurofins Discoverx Supplier No. 930813E3CP19L
Add to Cart
Add to Cart
Request a Quote
The PathHunter eXpress TORC2 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the CRTC/CREB pathway. This kit assesses ligand (e.g. forskolin) based pathway activation via detection of resulting TORC2 (CRTC2) nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged TORC2 (CRTC2) and nuclear localized EA. Upon pathway activation, induced nuclear translocation of TORC2 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.