Learn More
Eurofins Discoverx PathHunter eXpress TRHR Total Internalization Assay, 2pl Kit. (93-0949E3CP5M)
Catalog No. 502001645
Encompass
The PathHunter TRHR Total Internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of TRHR via detection of localization to the early endosome. The included cell line overexpresses EA-tagged TRHR and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the TRHR receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1645 Supplier Eurofins Discoverx Supplier No. 930949E3CP5M
The PathHunter TRHR Total Internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of TRHR via detection of localization to the early endosome. The included cell line overexpresses EA-tagged TRHR and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the TRHR receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.