Learn More
Eurofins Discoverx cAMP Hunter eXpress VIPR1 CHO-K1 GPCR Assay, 10-Plate Kit. (95-0093E2CP2L)
Catalog No. 502002952
Encompass
The cAMP Hunter VIPR1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. VIP) based activation of VIPR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type VIPR1, a naturally Gs coupled GPCR. Upon activation of VIPR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2952 Supplier Eurofins Discoverx Supplier No. 950093E2CP2L
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter VIPR1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. VIP) based activation of VIPR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type VIPR1, a naturally Gs coupled GPCR. Upon activation of VIPR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.