Learn More
Eurofins Discoverx PathHunter eXpress XBP1 Nuclear Translocation Assay, 10pl Kit. (93-0857E3CP5L)
Catalog No. 502001432
Encompass
The PathHunter eXpress XBP1 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the IRE1/XBP1 pathway. This kit assesses ligand (e.g. thapsigargin) based pathway activation via detection of resulting XBP1 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged XBP1 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of XBP1 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1432 Supplier Eurofins Discoverx Supplier No. 930857E3CP5L
Add to Cart
Add to Cart
Request a Quote
The PathHunter eXpress XBP1 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the IRE1/XBP1 pathway. This kit assesses ligand (e.g. thapsigargin) based pathway activation via detection of resulting XBP1 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged XBP1 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of XBP1 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.