Learn More
MSC Tiger-Vac Explosion-Proof HEPA Cleaner, Electric, 6.6 gal Capacity, Stainless Steel Tank
Catalog No. 502838281
Encompass
This explosion-proof and HEPA cleaner features an electric power source and a HEPA filter. It has a 6.6 gal capacity and a stainless steel tank, operating at 1.3 hp and 9A.
  • Electric power source for reliable operation
  • HEPA filtration ensures effective air purification
  • 6.6 gal tank capacity for ample collection
  • Constructed with a durable stainless steel tank
  • Maximum airflow of 132 CFM for efficient cleaning
  • Operates at 1.3 hp and 9A for powerful performance
  • Sound level of 85.0 dB
  • CSA certified for Class I, Division 1, Division 2, Group D, T3C & Division 1, Class II, Groups E, F & G
Catalog No. 50-283-8281 Supplier MSC Supplier No. 61162566
Add to Cart
Add to Cart
Request a Quote
This explosion-proof and HEPA cleaner features an electric power source and a HEPA filter. It has a 6.6 gal capacity and a stainless steel tank, operating at 1.3 hp and 9A.
  • Electric power source for reliable operation
  • HEPA filtration ensures effective air purification
  • 6.6 gal tank capacity for ample collection
  • Constructed with a durable stainless steel tank
  • Maximum airflow of 132 CFM for efficient cleaning
  • Operates at 1.3 hp and 9A for powerful performance
  • Sound level of 85.0 dB
  • CSA certified for Class I, Division 1, Division 2, Group D, T3C & Division 1, Class II, Groups E, F & G