Learn More
MSC Tiger-Vac Explosion-Proof & HEPA Cleaner, Electric, HEPA Filter, 3.4 gal Capacity
Catalog No. 502838133
Encompass
The Tiger-Vac Explosion-Proof & HEPA Cleaner is an electric vacuum designed for hazardous environments. It features a HEPA filtration system and a durable stainless steel tank, offering powerful cleaning with a 1.3 hp motor and a 3.4-gallon capacity. This unit is CSA certified for various hazardous classifications.
  • Electric power source for reliable operation
  • HEPA filtration for critical dust and particulate control
  • 3.4 gal capacity for efficient collection
  • 1.3 hp motor provides strong suction
  • Constructed with a stainless steel tank for durability
  • CSA Certified for Class I, Division 1, Division 2, Group D, T3C & Division 1, Class II, Groups E, F & G
  • Maximum air flow of 132 CFM
  • Operates at a sound level of 85.0 dB
Catalog No. 50-283-8133 Supplier MSC Supplier No. 49025034
Add to Cart
Add to Cart
Request a Quote
The Tiger-Vac Explosion-Proof & HEPA Cleaner is an electric vacuum designed for hazardous environments. It features a HEPA filtration system and a durable stainless steel tank, offering powerful cleaning with a 1.3 hp motor and a 3.4-gallon capacity. This unit is CSA certified for various hazardous classifications.
  • Electric power source for reliable operation
  • HEPA filtration for critical dust and particulate control
  • 3.4 gal capacity for efficient collection
  • 1.3 hp motor provides strong suction
  • Constructed with a stainless steel tank for durability
  • CSA Certified for Class I, Division 1, Division 2, Group D, T3C & Division 1, Class II, Groups E, F & G
  • Maximum air flow of 132 CFM
  • Operates at a sound level of 85.0 dB