Learn More
Optimize Technologies EXP™2 10-32 TI-LOK Fitting for all 1/16" OD Tubing, 10pk (10 EXP™2 TI-LOK fittings & 1 EXP™2 Driver)
SDP
Catalog No. 502006569
Encompass_Preferred
The EXP™2 TI-LOK Fittings offer capillary HPLC and UHPLC connections rated to 20,000+ psi in any port and easily fit into tight spaces such as injection valves, column couplers, sample loops and column ovens. The Hand-Tight fittings allow users the versatility to change out capillary tube ID, length or end fittings to create any tube set configuration. Available with 10-32 threads for 1/16" ports and 6-32 and 6-40 threads for 1/32” ports. EXP™2 TI-LOK Fittings feature built-in PEEK ferrules and threaded nuts precision machined from titanium. The ferrule does not permanently swage to the tubing, allowing for many repeat connections and the ability to adjust the tubing to different port depths. EXP™2 TI-LOK Fittings are compatible with PEEK, PEEKsil, Hastelloy C and stainless steel tubing. When used with the PEEKsil the fittings create a zero-dead-volume (ZDV) connection without fracturing the fused silica.
Catalog No. 50-200-6569 Supplier Optimize Technologies Supplier No. 152405205
The EXP™2 TI-LOK Fittings offer capillary HPLC and UHPLC connections rated to 20,000+ psi in any port and easily fit into tight spaces such as injection valves, column couplers, sample loops and column ovens. The Hand-Tight fittings allow users the versatility to change out capillary tube ID, length or end fittings to create any tube set configuration. Available with 10-32 threads for 1/16" ports and 6-32 and 6-40 threads for 1/32” ports. EXP™2 TI-LOK Fittings feature built-in PEEK ferrules and threaded nuts precision machined from titanium. The ferrule does not permanently swage to the tubing, allowing for many repeat connections and the ability to adjust the tubing to different port depths. EXP™2 TI-LOK Fittings are compatible with PEEK, PEEKsil, Hastelloy C and stainless steel tubing. When used with the PEEKsil the fittings create a zero-dead-volume (ZDV) connection without fracturing the fused silica.