Learn More
Optimize Technologies EXP™2 6-32 Fitting Kit for 1/32" OD SS Tubing (2 Titanium Hybrid Ferrules, 2 EXP™2 Nuts & 1 EXP™2 Driver)
SDP
Catalog No. 502006559
Encompass_Preferred
Featuring a small 3/16” hex-head EXP™2 nut, EXP™ Original Titanium Hybrid Ferrule and the new EXP™2 Driver, the EXP™2 Fittings offer hand-tight connections rated to 20,000 psi+. The EXP™2 Nuts are available with 6-32, 6-40 and 10-32 threads and have been scaled down to allow for maximum working room in cramped spaces. The EXP™2 Driver is a small, easy to use knurled torque driver which snaps on to the nuts for hand-tightening and loosening. It is also slotted so that it can be easily removed from the nut and tubing after use. The Hybrid Ferrule provides a perfect seal with every connection, yet the swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides unlimited value through repeated zero-dead-volume connections with any port configuration. EXP™2 fittings are the premier adjustable nut and ferrule compression system for extreme high-pressure connections.
Catalog No. 50-200-6559 Supplier Optimize Technologies Supplier No. 151904958
Featuring a small 3/16” hex-head EXP™2 nut, EXP™ Original Titanium Hybrid Ferrule and the new EXP™2 Driver, the EXP™2 Fittings offer hand-tight connections rated to 20,000 psi+. The EXP™2 Nuts are available with 6-32, 6-40 and 10-32 threads and have been scaled down to allow for maximum working room in cramped spaces. The EXP™2 Driver is a small, easy to use knurled torque driver which snaps on to the nuts for hand-tightening and loosening. It is also slotted so that it can be easily removed from the nut and tubing after use. The Hybrid Ferrule provides a perfect seal with every connection, yet the swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides unlimited value through repeated zero-dead-volume connections with any port configuration. EXP™2 fittings are the premier adjustable nut and ferrule compression system for extreme high-pressure connections.