Learn More
Optimize Technologies EXP™2 Filter Cartridge, 3mm, 0.2µm, 10/pk
SDP
Supplier:  Optimize Technologies 150405264
Encompass_Preferred
The EXP™2 Pre-Column and In-Line Hand-Tight Filters are ideal for protecting small-particle UHPLC columns and systems at extreme pressures. Such techniques analyze samples in the most demanding applications, decreasing the life of expensive hardware. EXP™2 Filters help extend the life of a UHPLC system and protect your investment without sacrificing performance. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet. EXP™2 Pre-Column Filter holders are available with Titanium Hybrid Ferrules for easy direct connection to any 10-32 port. The filter holder is a complete package and includes fittings to provide repeated zero-dead-volume connections. Use with holder part number 15-04-05242 or 15-04-05258.
Catalog No. 50-200-6496