Learn More
Medchemexpress LLC EXP3179 10mM 1mL | 114798-36-6 | 420.89 | C22H21ClN6O | 1 ML
SDP
Catalog No. 5000307560
Encompass_Preferred
EXP3179 is an important intermediate aldehyde metabolite of Losartan EXP3179 has no AT1-R blocking activity but potently inhibits the expression of endothelial cyclooxygenase (COX)-2 EXP3179 exerts potent anti-inflammatory actions[1]
Catalog No. 50-003-07560 Supplier Medchemexpress LLC Supplier No. HY11495010MM 1ML
EXP3179 is an important intermediate aldehyde metabolite of Losartan EXP3179 has no AT1-R blocking activity but potently inhibits the expression of endothelial cyclooxygenase (COX)-2 EXP3179 exerts potent anti-inflammatory actions[1]