Learn More
MSC Sealed Air Instapak Quick RT, 10, 15x18 in, 36/Case
Catalog No. 501919426
Encompass
Instapak Quick RT on-demand foam bags provide a portable, room temperature foam-in-bag system for protective packaging. Designed for shipping, these expandable foam bags deliver instant, custom-fit cushioning without the need for specialized equipment or preheating.
  • Provides easy-to-use, instant protective foam cushions
  • Portable system does not require a preheating warming unit
  • Chemicals activated inside the bag, eliminating cleanup
  • Foam expands up to 27 times its original size in seconds
  • Simple press, pat, and pack activation technique requires no equipment or training
  • Bags can be returned for landfill diversion
  • Recommended operating temperature range of 65–90° F
  • Recommended for an 8 × 8 × 8 in box
Catalog No. 50-191-9426 Supplier MSC Supplier No. 39590419
Instapak Quick RT on-demand foam bags provide a portable, room temperature foam-in-bag system for protective packaging. Designed for shipping, these expandable foam bags deliver instant, custom-fit cushioning without the need for specialized equipment or preheating.
  • Provides easy-to-use, instant protective foam cushions
  • Portable system does not require a preheating warming unit
  • Chemicals activated inside the bag, eliminating cleanup
  • Foam expands up to 27 times its original size in seconds
  • Simple press, pat, and pack activation technique requires no equipment or training
  • Bags can be returned for landfill diversion
  • Recommended operating temperature range of 65–90° F
  • Recommended for an 8 × 8 × 8 in box